missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FABP2/I-FABP Polyclonal specifically detects FABP2/I-FABP in Human, Mouse, Fish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
Specifications
| Applications | Western Blot, Immunohistochemistry (Paraffin), ELISA |
| Classification | Polyclonal |
| Dilution | Western Blot 0.001 ug/ml, Immunohistochemistry 2 - 3 ug/ml, Immunohistochemistry-Paraffin 2-3 ug/ml, Peptide ELISA Detection limit 1:64000 |
| Gene Alias | FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein |
| Molecular Weight of Antigen | 15 kDa |
| Purification Method | Affinity Purified |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?