missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP3/H-FABP Antibody (rFABP3/8534), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21008H
This item is not returnable.
View return policy
Description
FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FABP3/H-FABP | |
| Monoclonal | |
| HRP | |
| FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
| Human recombinant FABP3/H-FABP protein fragment (around aa1-127) (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism | |
| 2170 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rFABP3/8534 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction