missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP3 Mouse anti-Human, Clone: OTI7C7, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814071S
This item is not returnable.
View return policy
Description
FABP3 Monoclonal antibody specifically detects FABP3 in Human samples. It is validated for ELISASpecifications
| FABP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05413 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2170 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI7C7 | |
| Unconjugated | |
| FABP3 | |
| 422 protein; Cardiac Fatty Acid Binding Protein; FABP 3; FABP11; Fabp3; Fabph1; Fabph-1; Fabph4; Fabph-4; fatty acid binding protein 11; Fatty acid binding protein 11 (FABP11); fatty acid binding protein 3; Fatty acid binding protein 3 heart; fatty acid binding protein 3, muscle and heart; Fatty acid binding protein heart; fatty acid binding protein heart 1; fatty acid binding protein heart 4; fatty acid-binding protein 3; Fatty acid-binding protein, heart; heart FABP; heart fatty acid binding protein; Heart type fatty acid binding protein; heart-type fatty acid-binding protein; H-FABP; M FABP; Mammary derived growth inhibitor; Mammary-derived growth inhibitor; MDGI; M-FABP; muscle and heart (mammary derived growth inhibitor); Muscle fatty acid binding protein; Muscle fatty acid-binding protein; Muscle O-FABP; Mylein protein P2 homolog / adopocyte; O-FABP | |
| Full length human recombinant protein of human FABP3 (NP_004093) produced in EXPI-293F cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction