missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FABP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586815
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
FABP4 belongs to the fatty acid binding protein (FABP) family. FABPs contribute to the transport of fatty acids and other lipids in various cellular pathways. FABP4 is expressed in adipocytes, macrophages, monocyte-derived dendritic cells, and myoepithelial cells. It delivers long-chain fatty acids and retinoic acid to their cognate receptors in the nucleus.
Specifications
| FABP4 | |
| Polyclonal | |
| Unconjugated | |
| FABP4 | |
| 3T3-L1 lipid-binding protein; 422/aP2; adipocyte fatty acid binding protein; Adipocyte lipid binding protein; adipocyte lipid-binding protein; adipocyte protein aP2; adipocyte-type fatty acid-binding protein; AFABP; A-FABP; ALBP; ALBP/Ap2; Ap2; epididymis secretory protein Li 104; FABP4; fatty acid binding protein 4; Fatty acid binding protein 4 adipocyte; fatty acid binding protein 4, adipocyte; fatty acid-binding protein 4; Fatty acid-binding protein, adipocyte; HEL S 104; HEL-S-104; Lbpl; myelin P2 protein homolog; P15; P2 adipocyte protein; Protein 422 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2167, 79451 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P15090, P70623 | |
| FABP4 | |
| A synthetic peptide corresponding to residues in Human FABP4. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction