missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP5/E-FABP Antibody (311215), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3077CL1
This item is not returnable.
View return policy
Description
FABP5/E-FABP Monoclonal antibody specifically detects FABP5/E-FABP in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF,Gene Knock-Out
Specifications
| FABP5/E-FABP | |
| Monoclonal | |
| CoraFluor 1 | |
| EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
| E. coli-derived recombinant human FABP5/E-FABP, Ala2-Glu135, Accession # Q01469 | |
| 0.1 mL | |
| Stem Cell Markers | |
| 2171 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF, Gene Knock-Out | |
| 311215 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction