missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP5/E-FABP Antibody (FABP5/3750), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79929FR
This item is not returnable.
View return policy
Description
FABP5/E-FABP Monoclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FABP5/E-FABP | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FABP5 | |
| Recombinant full-length human FABP5/E-FABP protein (Uniprot: Q01469) | |
| 0.1 mL | |
| Stem Cell Markers | |
| Human | |
| IgG2a κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FABP5/3750 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction