missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 227.00 - € 326.00
Specifications
| Antigen | FABP6 |
|---|---|
| Dilution | Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611176
|
Novus Biologicals
NBP2-99577-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606965
|
Novus Biologicals
NBP2-99577-50ul |
50 μg |
€ 227.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP6 Polyclonal antibody specifically detects FABP6 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| FABP6 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| PBS (pH 7.0) | |
| 2172 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| fatty acid binding protein 6, ileal, Fatty acid-binding protein 6, gastrotropin, I-15PIntestinal 15 kDa protein, I-BABPIntestinal bile acid-binding protein, I-BALB, I-BAP, ILBP3, ILBPGT, ileal bile acid binding protein, ILLBPIleal lipid-binding protein, illeal lipid-binding protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse FABP6. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title