missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP7/B-FABP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16399
This item is not returnable.
View return policy
Description
FABP7/B-FABP Polyclonal specifically detects FABP7/B-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| FABP7/B-FABP | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| B-FABPDKFZp547J2313, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related | |
| Rabbit | |
| 15 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication, Stem Cells | |
| 2173 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 | |
| O15540 | |
| FABP7 | |
| Full length human FABP7 Recombinant protein. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (87%), Rat (88%), Bovine (95%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction