missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523469
This item is not returnable.
View return policy
Description
FABP7 was initially isolated from a human fetal brain cDNA library and whose mRNA was expressed in adult brain and muscle tissues at low levels. The protein encoded by this gene is a member of the fatty acid binding protein (FABPs) family, a group of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs are thought to play roles in fatty acid uptake, transport, and metabolism. FABP7 is a downstream gene of the Pax6 transcription factor and has been suggested to be essential for the maintenance of neuroepithelial cells during early cortical development. More recently, FABP7 was found to be frequently expressed in melanomas. Down-regulation of FABP7 through RNAi expression could reduce in vitro cell proliferation and Matrigel invasion, suggesting that FABP7 may be a potential target for the development of diagnostic and therapeutic tools.
Specifications
| FABP7 | |
| Polyclonal | |
| Unconjugated | |
| FABP7 | |
| B FABP; BFABP; B-FABP; BLBP; brain lipid binding protein; brain lipid-binding protein; Brain-type fatty acid-binding protein; DKFZp547J2313; FABP 7; FABP7; FABPB; fatty acid binding protein 7; fatty acid binding protein 7, brain; fatty acid-binding protein 7; Fatty acid-binding protein, brain; fatty acid-binding protein, brain-like protein; hypothetical protein DKFZp547J2313; Mammary-derived growth inhibitor related; mammary-derived growth inhibitor-related; MRG; OTTHUMP00000017119; RP11-548E19.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100067280, 12140, 2173, 777787 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O15540, P51880, Q09139 | |
| FABP7 | |
| A portion of amino acids 80-132 of human FABP7 | |
| 100 μg | |
| Primary | |
| Bovine, Equine, Human, Marsupial, Mouse, Non-human Primate | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction