missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP8/M-FABP/Myelin P2 Protein Antibody (3H3F4D5), Novus Biologicals™
Mouse Monoclonal Antibody
€ 231.00 - € 326.00
Specifications
| Antigen | FABP8/M-FABP/Myelin P2 Protein |
|---|---|
| Clone | 3H3F4D5 |
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618716
|
Novus Biologicals
NBP3-06650-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682418
|
Novus Biologicals
NBP3-06650-50ul |
50 μg |
€ 231.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP8/M-FABP/Myelin P2 Protein Monoclonal antibody specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for ELISASpecifications
| FABP8/M-FABP/Myelin P2 Protein | |
| ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PMP2 (Accession#: NP_002668.1; Ser 2-Val 132). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3H3F4D5 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 5375 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title