missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FABP9 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5119106

 View more versions of this product

Product Code. 17780514

  • € 449.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FABP9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Fatty acid-binding protein 9 (FABP9), also known as testis FABP, is a gene located on chromosome 8q21.1 that encodes a protein primarily expressed in the testis, where it plays a critical role in fatty acid metabolism and transport. FABP9 is part of a larger family of fatty acid-binding proteins that facilitate the intracellular movement and storage of lipids, impacting cellular signaling and energy homeostasis. Structurally, FABP9 is adapted to bind long-chain fatty acids and other hydrophobic molecules, contributing to the regulation of lipid dynamics within spermatogenic cells. The expression of FABP9 is crucial during spermatogenesis, influencing sperm maturation and fertility. Beyond its reproductive role, variations in FABP9 have been studied in relation to metabolic disorders, linking its lipid regulatory functions to broader physiological processes. Research continues to explore FABP9's potential involvement in systemic lipid metabolism and its implications for diseases associated with lipid dysregulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FABP9
Polyclonal
Unconjugated
Fabp9
15 kDa perforatorial protein; 1700007P10Rik; FABP9; FABP-9; fatty acid binding protein 9; fatty acid binding protein 9, testis; fatty acid-binding protein 9; lipid-binding protein; PERF; PERF 15; Perf15; testis fatty acid binding protein; testis lipid binding protein; testis lipid-binding protein; Testis-type fatty acid-binding protein; T-FABP; TLBP
Rabbit
Antigen affinity chromatography, Protein A
RUO
21884, 64822
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
O08716, P55054
Fabp9
Recombinant protein Fatty Acid Binding Protein 9, Testis. The antigen corresponds to amino acid range 1-132 of the target protein.
200 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.