missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FACA/FANCA Antibody (SR1233), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | FACA/FANCA |
|---|---|
| Clone | SR1233 |
| Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 1:40 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627734
|
Novus Biologicals
NBP3-22105-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639784
|
Novus Biologicals
NBP3-22105-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FACA/FANCA Monoclonal antibody specifically detects FACA/FANCA in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| FACA/FANCA | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1:40 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FA, FA1, FAAMGC75158, FACAFanconi anemia group A protein, FAH, FA-H, FANCH, Fanconi anemia, complementation group A, Fanconi anemia, complementation group H, Fanconi anemia, type 1, Protein FACA | |
| A synthesized peptide derived from human FACA/FANCA (Uniprot #: O15360) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1233 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2175 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title