missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor D Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
€ 134.00 - € 455.00
Specifications
| Antigen | Factor D |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17684326
|
Invitrogen™
MA543874 |
20 μL |
€ 134.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17674326
![]() |
Invitrogen™
MA543873 |
100 μL |
€ 455.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.Specifications
| Factor D | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 28 kDa adipocyte protein; adipsin; Adn; C3 convertase activator; Cfd; Complement factor D; complement factor D (adipsin); complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Df; EC 3.4.21.46; Endogenous vascular elastase; EVE; PFD; Properdin factor D | |
| CFD | |
| IgG2a κ | |
| Protein A/G | |
| Antibody |
| C1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P00746 | |
| 1675 | |
| Recombinant CFD (Leu7-Asp249) expressed in E.coli. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CFD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title