missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor IX Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98269AF700
This item is not returnable.
View return policy
Description
Factor IX Polyclonal specifically detects Factor IX in Mouse samples. It is validated for ELISA.
Specifications
| Factor IX | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2158 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 700 | |
| Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC | |
| Produced in rabbits immunized with purified, recombinant Factor IX (P16294; Met1-Thr471). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction