Learn More
Factor IX Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98269H
Product Code. 18083038
  • € 415.00 / 0.10mL
Description
Factor IX Polyclonal specifically detects Factor IX in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Factor IX
Polyclonal
PBS
Rabbit
Affinity purified
RUO
2158
Store at 4°C in the dark.
IgG
ELISA
HRP
Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC
Produced in rabbits immunized with purified, recombinant Factor IX (P16294; Met1-Thr471).
0.1 mL