missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor IX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99802-100ul
This item is not returnable.
View return policy
Description
Factor IX Polyclonal antibody specifically detects Factor IX in Human samples. It is validated for ELISA
Specifications
| Factor IX | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC | |
| Produced in rabbits immunized with purified, recombinant Human Factor IX (Accession#: AAB59620.1; Met1-Thr461) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2158 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction