missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor IX Monoclonal Antibody (CaFIX-1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517637
This item is not returnable.
View return policy
Description
MA5-17637 targets Factor IX/IXa in ELISA and WB applications and shows reactivity with Human samples. The MA5-17637 immunogen is human Factor IX (CaFIX-1) purified from plasma.
Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
Specifications
| Factor IX | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P00740 | |
| F9 | |
| Human Factor IX (CaFIX-1) purified from plasma. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| CaFIX-1 | |
| Unconjugated | |
| F9 | |
| AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8 | |
| Mouse | |
| Protein G | |
| RUO | |
| 2158 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction