missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor IX Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA143046
This item is not returnable.
View return policy
Description
PA1-43046 detects Factor IX in human samples. PA1-43046 has successfully been used in Western blot and ELISA applications. PA1-43046 was derived from human factor IX.
Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
Specifications
| Factor IX | |
| Polyclonal | |
| Unconjugated | |
| F9 | |
| AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8 | |
| Sheep | |
| SDS-PAGE | |
| RUO | |
| 2158 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 15 mg/mL | |
| 50% water with 50% glycerol and no preservative | |
| P00740 | |
| F9 | |
| Human Factor IX. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction