missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VII Monoclonal Antibody (RFF-VII/2)
Factor VII Monoclonal Antibody (RFF-VII/2)
Brand: Invitrogen MA516932
This item is not returnable.
View return policy
Description
Specifications
| Factor VII | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P08709 | |
| F7 | |
| Purified Factor VII. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Radioimmune Assays (RIA), Western Blot | |
| RFF-VII/2 | |
| Unconjugated | |
| F7 | |
| AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA | |
| Mouse | |
| Protein A | |
| RUO | |
| 2155 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction