missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VIII Antibody (2A5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62559H
This item is not returnable.
View return policy
Description
Factor VIII A2 domain Monoclonal specifically detects Factor VIII A2 domain in Human samples. It is validated for ELISA.
Specifications
| Factor VIII A2 domain | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| F8 | |
| Purified human factor VIII | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 2157 | |
| Human | |
| Purified |
| ELISA | |
| 2A5 | |
| ELISA | |
| AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB100-62559 recognizes the 83 kDa light chain of Factor VIII. It does not cross-react with Von Willebrand factor. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction