missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VIII Antibody (402), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90255C
This item is not returnable.
View return policy
Description
Factor VIII Monoclonal specifically detects Factor VIII in Human samples. It is validated for ELISA.
Specifications
| Factor VIII | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 402 | |
| ELISA | |
| AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Factor VIII protein. | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 2157 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction