missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VIII Antibody (RFF-VIII:c/8), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50398DL594
This item is not returnable.
View return policy
Description
Factor VIII Monoclonal antibody specifically detects Factor VIII in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Factor VIII | |
| Monoclonal | |
| DyLight 594 | |
| AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component | |
| Purified human factor VIII | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 2157 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunofluorescence | |
| RFF-VIII:c/8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction