missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor VIII:c heavy chain Antibody (RFF-VIII:c/10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53119MFV500
This item is not returnable.
View return policy
Description
Factor VIII:c heavy chain Monoclonal antibody specifically detects Factor VIII:c heavy chain in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Factor VIII:c heavy chain | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Purified human factor VIII | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunocytochemistry | |
| RFF-VIII:c/10 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction