missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor VIII Recombinant Rabbit Monoclonal Antibody (402), HRP, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 163.00 - € 510.00
Specifications
| Antigen | Factor VIII |
|---|---|
| Clone | 402 |
| Concentration | 0.65 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019484
![]() |
Invitrogen™
MA552346 |
100 μg |
€ 510.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019295
|
Invitrogen™
MA552347 |
20 μg |
€ 163.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Factor VIII Recombinant Monoclonal antibody specifically detects Factor VIII in Human samples. It is validated for ELISASpecifications
| Factor VIII | |
| 0.65 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| AHF; Antihemophilic factor; CF8; Cf-8; coagulation factor VIII; coagulation factor VIII A1 domain; coagulation factor VIII C2 domain; coagulation factor VIII, procoagulant component; coagulation factor VIII, procoagulant component (hemophilia A); coagulation factor VIIIc; DXS1253E; F8; F8B; F8C; Factor 8; Factor VIII; Factor VIII B chain; factor VIII F8B; factor VIII precursor; Factor VIIIa heavy chain, 200 kDa isoform; Factor VIIIa heavy chain, 92 kDa isoform; Factor VIIIa light chain; factor8; FVIII; HEMA; Procoagulant component; RP11-115M6.7 | |
| Recombinant Human Coagulation Factor VIII/FVIII/F8 protein. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| F8 |
| 402 | |
| ELISA | |
| HRP | |
| Rabbit | |
| Human | |
| P00451 | |
| 2157 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title