missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XII heavy chain Antibody (10-11-5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41375PCP
This item is not returnable.
View return policy
Description
Factor XII heavy chain Monoclonal antibody specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| Factor XII heavy chain | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2161 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, ELISA | |
| 2005-10-11 00:00:00 | |
| Western Blot, ELISA, Sandwich ELISA | |
| beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor | |
| Affinity purified human factor XII. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction