missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XII heavy chain Antibody (OTI1H5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70695B
This item is not returnable.
View return policy
Description
Factor XII heavy chain Monoclonal specifically detects Factor XII heavy chain in Human samples. It is validated for Immunohistochemistry.
Specifications
| Factor XII heavy chain | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| F12 | |
| Recombinant protein fragment corresponding to amino acids 73-372 of human Factor XII heavy chain (NP_000496) produced in E.coli. | |
| 0.1 mL | |
| 2161.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI1H5 | |
| Immunohistochemistry | |
| beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction