Learn More
Factor XII Mouse anti-Human, CoraLite 488, Clone: 2E2F11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66089-100UL
Description
The Factor XII antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Factor XII. This antibody recognizes human antigen. The Factor XII antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| Factor XII | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence | |
| 2E2F11 | |
| CoraLite 488 | |
| 100 μL | |
| 2161 | |
| IgG2b |