Learn More
Factor XII Mouse anti-Human, CoraLite 488, Clone: 2E2F11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66089-100UL
Product Code. 16860716
  • € 450.00 / 100µL
Description
The Factor XII antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human Factor XII. This antibody recognizes human antigen. The Factor XII antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
Factor XII
Monoclonal
2000 μg/mL
Mouse
Primary
Human
Immunofluorescence
2E2F11
CoraLite 488
100 μL
2161
IgG2b