missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XIIIa Mouse, Clone: F13A1/1683, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2162-MSM3-P0
This item is not returnable.
View return policy
Specifications
| Factor XIIIa | |
| Cytoplasmic and Secreted | |
| F13A1/1683 | |
| Unconjugated | |
| NP_000120 | |
| Mouse | |
| 83kDa | |
| Protein A/G Purified | |
| RUO | |
| 2162 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| F13A1 | |
| Recombinant fragment of human Factor XIIIa protein (aa46-181) | |
| 293T Cells. Placenta. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction