missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XIIIb Antibody (OTI1A3), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70697AF750
This item is not returnable.
View return policy
Description
Factor XIIIb Monoclonal specifically detects Factor XIIIb in Human samples. It is validated for Western Blot.
Specifications
| Factor XIIIb | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| F13B | |
| Recombinant protein fragment corresponding to amino acids 357-661 of human Factor XIII (NP_001985) produced in E.coli. | |
| 0.1 mL | |
| 2165.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1A3 | |
| Western Blot | |
| coagulation factor XIII B chain, coagulation factor XIII, B polypeptide, F13B, fibrin-stabilizing factor B subunit, FXIIIB, protein-glutamine gamma-glutamyltransferase B chain, TGase, transglutaminase B chain | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction