Learn More
Memmert™ Factory Calibration (3 Temperatures) for IN110
Product Code. 17577355 Shop All Memmert Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
17577355 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17577355 Supplier Memmert™ Supplier No. IN110D00126
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Calibration at Memmert factory for three temperature value.
  • For three temperature values: 37°C, 52°C, and 70°C
Specifications
For Use With (Equipment) IN110 Incubator
Description Measuring point chamber center
Calibration Type Certificate for 3 temperatures
Product Type Standard Works Calibration Certificate