Learn More
Fadrozole hydrochloride, MedChemExpress
Product Code. 16478617
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16478617
Brand: Medchem Express HY14247
MedChemExpress Fadrozole hydrochloride (CGS 16949A) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.
Fadrozole hydrochloride (CGS 16949A) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.
Specifications
Chemical Name or Material Fadrozole hydrochloride
CAS 102676-31-3
Color White
Purity Grade Notes Research
Molecular Formula C14H14ClN3
Quantity 5 mg
Synonym CGS 16949A(Rac)-FAD286 hydrochloride
Solubility Information DMSO : 100 mg/mL (385.02 mM; Need ultrasonic and warming) ∣H2O : 100 mg/mL (385.02 mM; Need ultrasonic)
SMILES N#CC1=CC=C(C2CCCC3=CN=CN23)C=C1.Cl
Molecular Weight (g/mol) 259.73
Formula Weight 259.73
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only