missing translation for 'onlineSavingsMsg'
Learn More

FADS2 Polyclonal Antibody
GREENER_CHOICE

FADS2 Polyclonal Antibody

Brand:  Invitrogen PA552336

Product Code. 15710291

  • € 560.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FADS2 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY
Specifications

Specifications

FADS2
Polyclonal
Unconjugated
Fads2
2900042M13Rik; Acyl-CoA 6-desaturase; D6D; delta(6) desaturase; delta(6) fatty acid desaturase; delta-6 desaturase; delta-6 fatty acid desaturase; delta-6-desaturase; DES6; FADS2; Fadsd2; FADSD6; Fatty acid desaturase 2; linoleoyl-CoA desaturase (delta-6-desaturase)-like 2; LLCDL2; SLL0262; TU13
Rabbit
Antigen affinity chromatography
RUO
9415
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O95864
Fads2
Recombinant protein corresponding to Human FADS2. Recombinant protein control fragment (Product #RP-88967).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.