missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31651
This item is not returnable.
View return policy
Description
FAF1 Polyclonal specifically detects FAF1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| FAF1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
| FAF1 | |
| Recombinant protein encompassing a sequence within the center region of human FAF1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.66mg/mL | |
| Western Blot 1:500-1:3000 | |
| CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1 | |
| Rabbit | |
| 74 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 11124 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction