missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF1 Antibody (OTI10D1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71679F
This item is not returnable.
View return policy
Description
FAF1 Monoclonal antibody specifically detects FAF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| FAF1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI10D1 | |
| Western Blot, Immunohistochemistry | |
| CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1 | |
| Human recombinant protein fragment corresponding to amino acids 415-650 of human FAF1 (NP_008982) produced in E.coli. | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 11124 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction