missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAC2 Monoclonal Antibody (OTI6G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810460
This item is not returnable.
View return policy
Description
PLAC2 (placenta-specific protein 2) is a 151 amino acid protein that is encoded by a gene located on human chromosome 19p13. Based on its amino acid sequence, PLAC2 is predicted to be a multi-pass membrane protein. There are no reported studies or functions reported for this protein.Specifications
| PLAC2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| LINC00036, NCRNA00036, PLAC2, onco-lncRNA-16 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 257000 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI6G4 | |
| Unconjugated | |
| TINCR | |
| TINCR | |
| Full length human recombit protein of human PLAC2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction