missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF1 Monoclonal Antibody (OTI5G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804140
This item is not returnable.
View return policy
Description
Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain.Specifications
| FAF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UNN5 | |
| FAF1 | |
| Human recombit protein fragment corresponding to amino acids 415-650 of human FAF1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5G9 | |
| Unconjugated | |
| FAF1 | |
| CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11124 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction