Learn More
Fagomine, MedChemExpress
Product Code. 16468187
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16468187
Brand: Medchem Express HY13005
MedChemExpress Fagomine is a mild glycosidase inhibitor. The Ki of the iminosugar Fagomine is 4.8 μM, 39 μM, and 70 μM for Amyloglucosidase (A.niger), β-Glucosidase (bovine), and Isomaltase (yeast), respectively.
Fagomine is a mild glycosidase inhibitor. The Ki of the iminosugar Fagomine is 4.8 μM, 39 μM, and 70 μM for Amyloglucosidase (A.niger), β-Glucosidase (bovine), and Isomaltase (yeast), respectively.
Specifications
Chemical Name or Material Fagomine
Physical Form Solid
CAS 53185-12-9
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C6H13NO3
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym D-Fagomine
Solubility Information H2O : ≥ 36 mg/mL (244.62 mM)
SMILES O[C@@H]1CCN[C@@H]([C@H]1O)CO
Molecular Weight (g/mol) 147.17
Formula Weight 147.17
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only