missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAHD2A Antibody (OTI6D9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71886AF532
This item is not returnable.
View return policy
Description
FAHD2A Monoclonal specifically detects FAHD2A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| FAHD2A | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FAHD2A | |
| Full length recombinant protein of human FAHD2A (NP_057128) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6D9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CGI-105, EC 3.-, fumarylacetoacetate hydrolase domain containing 1, fumarylacetoacetate hydrolase domain containing 2A, fumarylacetoacetate hydrolase domain-containing protein 2A, MGC131995 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 51011.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction