missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAHD2A Monoclonal Antibody (OTI6D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525048
This item is not returnable.
View return policy
Description
FAHD2A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
May have hydrolase activity.
Specifications
| FAHD2A | |
| Monoclonal | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96GK7 | |
| FAHD2A | |
| Full length human recombinant protein of FAHD2A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI6D9 | |
| Unconjugated | |
| FAHD2A | |
| 1500003K14Rik; B430104H02Rik; CGI-105; CGI-105 protein; Fahd2; FAHD2A; fumarylacetoacetate hydrolase domain containing 1; fumarylacetoacetate hydrolase domain containing 2A; fumarylacetoacetate hydrolase domain-containing protein 2; Fumarylacetoacetate hydrolase domain-containing protein 2A; LOC100062565; LOC100731203; RGD1563674 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51011 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction