Learn More
FAHD2A Mouse anti-Human, Clone: OTI9D4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500944
Beskrivelse
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May have hydrolase activity.Tekniske data
| FAHD2A | |
| Monoclonal | |
| Unconjugated | |
| FAHD2A | |
| CGI-105 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51011 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI9D4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96GK7 | |
| FAHD2A | |
| Full length human recombit protein of human FAHD2A produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.