Learn More
Invitrogen™ FAIM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529200
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (89%), Rat (90%), Xenopus laevis (86%), Pig (93%), Rhesus Monkey (96%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells.
Specifications
| FAIM | |
| Polyclonal | |
| Unconjugated | |
| FAIM | |
| Faim; FAIM 1; FAIM1; Fas apoptotic inhibitory molecule; fas apoptotic inhibitory molecule 1; rFAIM | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23873, 55179 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NVQ4, Q9WUD8 | |
| FAIM | |
| Full length human FAIM1 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.