missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAIM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583613
This item is not returnable.
View return policy
Description
Immunogen sequence: MLLPFIRTLP LLCYNHLLVS PDSATLSPPY SLEKMTDLVA VWDVALSDGV HKIEFEHGTT SGKRVVYVDG K Highest antigen sequence indentity to the following orthologs: Rat - 61%, Mouse - 62%.
Plays a role as an inducible effector molecule that mediates Fas resistance produced by surface Ig engagement in B cells.
Specifications
| FAIM | |
| Polyclonal | |
| Unconjugated | |
| FAIM | |
| Faim; FAIM 1; FAIM1; Fas apoptotic inhibitory molecule; fas apoptotic inhibitory molecule 1; rFAIM | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55179 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVQ4 | |
| FAIM | |
| Recombinant protein corresponding to Human FAIM. Recombinant protein control fragment (Product #RP-92062). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction