missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAIM3 Antibody (992338) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9494A
This item is not returnable.
View return policy
Description
FAIM3 Monoclonal antibody specifically detects FAIM3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| FAIM3 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 992338 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| Fas apoptotic inhibitory molecule 3, Fc mu receptor, FCMR, immunoglobulin mu Fc receptor, Regulator of Fas-induced apoptosis Toso, TOSOIgM Fc receptor | |
| Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3, Arg18-Gly251, Accession # O60667 | |
| 0.1 mL | |
| Apoptosis | |
| 9214 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction