missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAIM3 Antibody (992338) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9494JF669
This item is not returnable.
View return policy
Description
FAIM3 Monoclonal antibody specifically detects FAIM3 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| FAIM3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Fas apoptotic inhibitory molecule 3, Fc mu receptor, FCMR, immunoglobulin mu Fc receptor, Regulator of Fas-induced apoptosis Toso, TOSOIgM Fc receptor | |
| Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3, Arg18-Gly251, Accession # O60667 | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 992338 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis | |
| 9214 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction