Learn More
FAIM3 Antibody (992338), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB9494MFV500
Product Code. 18502229
  • € 652.00 / 0.10mL
Description
FAIM3 Monoclonal antibody specifically detects FAIM3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FAIM3
Monoclonal
0.1 mL
Western Blot, CyTOF