missing translation for 'onlineSavingsMsg'
Learn More

Logistik Unicorp™ Faithful Royal Blue Color 65% Polyester/35% Cotton Lab Coats

Catalog Number p-8006091
Click to view available options
Size (Metric) Chest:
92cm
96cm
100cm
104cm
132cm
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Manufactured in hardwearing polycotton. Logistik Unicorp™ Faithful Royal Blue Color 65% Polyester/35% Cotton Lab Coats offer durability and ease of movement.

  • Poly/cotton blend
  • Concealed stud front
  • One breast pocket
  • Two lower patch pockets
  • Center back vent 
  • 245gm
  • Not CE marked
Type Lab Coat
Size (Metric) Chest 100cm
For Use With (Application) Used for general laboratory safety
Includes One breast pocket, two lower patch pockets, centre back vent
Weight (Metric) 245 g/m2
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'