missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ FAK Inhibitor 14
GREENER_CHOICE

Product Code. 18074182
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18074182 10 mg 10mg
18064842 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074182 Supplier Tocris Bioscience™ Supplier No. 3414/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective FAK inhibitor

FAK Inhibitor 14 is a selective focal adhesion kinase (FAK) inhibitor that displays no significant activity at a range of other kinases including EGFR, PDGFR and IGF-RI. FAK Inhibitor 14 prevents FAK autophosphorylation at Y397 (IC50 = 1 μM), promotes cell detachment and inhibits cell adhesion in vitro. FAK Inhibitor 14 exhibits antiproliferative activity in a variety of human tumor cell lines in vitro and in breast cancer cells in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4506-66-5
Molecular Formula C6H14Cl4N4

Specifications

Quantity 10 mg
Formula Weight 284.01
Purity >98%
Molecular Weight (g/mol) 284.006
Chemical Name or Material FAK Inhibitor 14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.