missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAK, p Tyr397 Antibody (SR1077), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 373.00 - € 497.00
Specifications
| Antigen | FAK |
|---|---|
| Clone | SR1077 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669954
|
Novus Biologicals
NBP3-21534-100ul |
100 μg |
€ 497.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653034
|
Novus Biologicals
NBP3-21534-50ul |
50 μg |
€ 373.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAK Monoclonal antibody specifically detects FAK in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| FAK | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.10, FADK 1, FADKFAK-related non-kinase polypeptide, FAK1FRNK, FAKEC 2.7.10.2, focal adhesion kinase 1, pp125FAK, Protein-tyrosine kinase 2, PTK2 protein tyrosine kinase 2 | |
| A synthesized peptide derived from human phospho-FAK [Tyr397] (Uniprot #: Q05397) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR1077 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5747 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title