missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAKD3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97546MFV450
This item is not returnable.
View return policy
Description
FAKD3 Polyclonal antibody specifically detects FAKD3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FAKD3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| FAST kinase domain-containing protein 3, FAST kinase domains 3, FLJ23274, MGC142123, MGC5297 | |
| Produced in rabbits immunized with E. coli-derived Human FAKD3 fragment. | |
| 0.1 mL | |
| Protein Kinase | |
| 79072 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction