Learn More
Falcon™ Express™ Cordless Pipet-Aid™ Pipet Filler Replacement Parts
Product Code. 10516961 Shop All Corning Products
Change view
Click to view available options
Tissue Culture Nosepiece
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10516961 Tissue Culture Nosepiece
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10516961 Supplier Corning Supplier No. 357593
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Description Tissue Culture Nosepiece
Autoclavable Not Autoclavable
For Use With (Equipment) Falcon™ Express™ cordless Pipet-Aid™ pipet filler